Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001098241.1 | NP_001091711.1 | 2 - 3 | 275 | 72 |
NM_001130441.1 | NP_001123913.1 | 2 - 3 | 275 | 72 | |
XM_006230535.2 | XP_006230597.2 | 2 - 3 | 269 | 72 | |
GenBank mRNA | BC086608.1 | - | 2 - 3 | 271 | 72 |
BC099130.1 | - | 2 - 3 | 297 | 72 | |
CO567790.1 | - | 2 - 3 | 275 | 72 |