Assay Design



Gene Transcripts

Gene Symbol
Nkpd1
Entrez Gene ID
680440
Gene Name
NTPase, KAP family P-loop domain containing 1
Gene Aliases
-
Chromosome Location
Chr.1: 80424507 - 80434115 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001057808.5 XP_001057808.2 5 - 5 4426 60
XM_006228500.2 XP_006228562.1 4 - 4 3979 60

View Full Product Details