Assay Design



Gene Transcripts

Gene Symbol
Sertad3
Entrez Gene ID
499108
Gene Name
SERTA domain containing 3
Gene Aliases
-
Chromosome Location
Chr.1: 84280945 - 84284681 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017513.1 NP_001017513.1 1 - 2 69 131
GenBank mRNA BC091229.1 - 1 - 2 69 131

View Full Product Details