Assay Design



Gene Transcripts

Gene Symbol
Psmc4
Entrez Gene ID
117262
Gene Name
proteasome (prosome, macropain) 26S subunit, ATPase, 4
Gene Aliases
Tbp7
Chromosome Location
Chr.1: 84978220 - 84986536 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_057122.1 NP_476463.1 5 - 6 597 95
GenBank mRNA BC063145.1 - 5 - 6 607 95
D50695.1 - 5 - 6 597 95
FQ226113.1 - 5 - 6 606 95

View Full Product Details