Assay Design



Gene Transcripts

Gene Symbol
Aldh2
Entrez Gene ID
29539
Gene Name
aldehyde dehydrogenase 2 family (mitochondrial)
Gene Aliases
-
Chromosome Location
Chr.12: 40466425 - 40498813 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032416.1 NP_115792.1 4 - 5 474 57
XM_006249385.2 XP_006249447.1 4 - 5 538 57
GenBank mRNA AF529165.2 - 4 - 5 351 57
AY566467.1 - 4 - 5 418 57
AY566468.1 - 4 - 5 418 57
AY566469.1 - 4 - 5 418 57
BC062081.1 - 4 - 5 450 57
X14977.1 - 4 - 5 474 57

View Full Product Details