Assay Design



Gene Transcripts

Gene Symbol
Vps29
Entrez Gene ID
288666
Gene Name
vacuolar protein sorting 29 homolog (S. cerevisiae)
Gene Aliases
-
Chromosome Location
Chr.12: 39690069 - 39699127 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105932.1 NP_001099402.1 1 - 2 87 122
GenBank mRNA AB578913.1 - 1 - 2 68 122
AB578915.1 - 1 - 2 73 122
AB578916.1 - 1 - 2 67 122
AB578921.1 - 1 - 2 71 122

View Full Product Details