Assay Design



Gene Transcripts

Gene Symbol
Galnt9
Entrez Gene ID
304571
Gene Name
UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 9
Gene Aliases
-
Chromosome Location
Chr.12: 52106506 - 52187198 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001122644.1 NP_001116116.1 5 - 6 1275 127
XM_008769333.1 XP_008767555.1 5 - 6 1309 127
XM_008769334.1 XP_008767556.1 5 - 6 1274 127

View Full Product Details