Assay Design



Gene Transcripts

Gene Symbol
Galnt9
Entrez Gene ID
304571
Gene Name
UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 9
Gene Aliases
-
Chromosome Location
Chr.12: 52106506 - 52187198 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107151.2 NP_001100621.1 2 - 3 267 69
NM_001122644.1 NP_001116116.1 7 - 8 1579 69
XM_008769334.1 XP_008767556.1 7 - 8 1578 69

View Full Product Details