Assay Design



Gene Transcripts

Gene Symbol
Wbscr16
Entrez Gene ID
360796
Gene Name
Williams-Beuren syndrome chromosome region 16
Gene Aliases
-
Chromosome Location
Chr.12: 25561703 - 25592337 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108332.1 NP_001101802.1 10 - 11 1362 116
XM_006249183.2 XP_006249245.1 9 - 10 1514 116

View Full Product Details