Assay Design



Gene Transcripts

Gene Symbol
Sumf2
Entrez Gene ID
360800
Gene Name
sulfatase modifying factor 2
Gene Aliases
-
Chromosome Location
Chr.12: 30465025 - 30490683 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025125.2 NP_001020296.2 4 - 5 414 86
GenBank mRNA BC091176.1 - 4 - 5 420 86

View Full Product Details