Assay Design



Gene Transcripts

Gene Symbol
Cmklr1
Entrez Gene ID
60669
Gene Name
chemokine-like receptor 1
Gene Aliases
-
Chromosome Location
Chr.12: 48743555 - 48796582 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006249505.2 XP_006249567.1 1 - 2 370 68
GenBank mRNA CB707803.1 - 1 - 2 253 68

View Full Product Details