Assay Design
Gene Transcripts
- Gene Symbol
- Fbxo21
- Entrez Gene ID
-
360818
- Gene Name
- F-box protein 21
- Gene Aliases
-
-
- Chromosome Location
-
Chr.12: 44244457 - 44279044 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC158866.1 |
- |
2 - 3 |
382 |
64 |
FQ212189.1 |
- |
2 - 3 |
395 |
64 |
View Full Product Details