Assay Design



Gene Transcripts

Gene Symbol
Tfr2
Entrez Gene ID
288562
Gene Name
transferrin receptor 2
Gene Aliases
Trfr2
Chromosome Location
Chr.12: 22177382 - 22194333 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105916.1 NP_001099386.1 12 - 13 1563 74
XM_006249116.2 XP_006249178.1 11 - 12 1515 74
XM_006249117.1 XP_006249179.1 12 - 13 1531 74
XM_006249118.1 XP_006249180.1 12 - 13 1534 74
XM_008769125.1 XP_008767347.1 11 - 12 1644 74
XM_008769126.1 XP_008767348.1 11 - 12 1474 74
XM_008769127.1 XP_008767349.1 11 - 12 1515 74
XM_008769128.1 XP_008767350.1 11 - 12 1515 74
GenBank mRNA BC166451.1 - 11 - 12 1628 74

View Full Product Details