Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001107157.1 | NP_001100627.1 | 6 - 7 | 1459 | 156 |
XM_006249158.1 | XP_006249220.1 | 5 - 6 | 1405 | 156 | |
XM_008769137.1 | XP_008767359.1 | 5 - 6 | 1260 | 156 | |
GenBank mRNA | BC166738.1 | - | 5 - 6 | 1346 | 156 |