Assay Design



Gene Transcripts

Gene Symbol
Tnfsf13b
Entrez Gene ID
498666
Gene Name
tumor necrosis factor (ligand) superfamily, member 13b
Gene Aliases
RGD1561519
Chromosome Location
Chr.16: 85275690 - 85306412 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109112.1 NP_001102582.1 2 - 3 748 79
XM_006253480.2 XP_006253542.1 2 - 3 1072 79
XM_006253481.2 XP_006253543.1 2 - 3 1072 79

View Full Product Details