Assay Design



Gene Transcripts

Gene Symbol
Tpte2
Entrez Gene ID
364629
Gene Name
transmembrane phosphoinositide 3-phosphatase and tensin homolog 2
Gene Aliases
Tpte
Chromosome Location
Chr.16: 74558529 - 74662194 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108877.1 NP_001102347.1 17 - 18 1472 70

View Full Product Details