Assay Design



Gene Transcripts

Gene Symbol
Rfwd2
Entrez Gene ID
360860
Gene Name
ring finger and WD repeat domain 2, E3 ubiquitin protein ligase
Gene Aliases
Cop1, RGD1304773
Chromosome Location
Chr.13: 76942933 - 77076015 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025126.1 NP_001020297.1 2 - 3 313 141
XM_006250113.2 XP_006250175.1 9 - 10 1512 141
XM_006250114.2 XP_006250176.1 9 - 10 1500 141
XM_006250115.2 XP_006250177.1 8 - 9 1452 141
XM_006250116.2 XP_006250178.1 8 - 9 1440 141
GenBank mRNA BC091284.1 - 2 - 3 313 141

View Full Product Details