Assay Design



Gene Transcripts

Gene Symbol
Klhl20
Entrez Gene ID
304920
Gene Name
kelch-like family member 20
Gene Aliases
RGD1309490
Chromosome Location
Chr.13: 78912361 - 78957222 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107192.1 NP_001100662.1 12 - 13 2493 98

View Full Product Details