Assay Design



Gene Transcripts

Gene Symbol
Slc41a1
Entrez Gene ID
363985
Gene Name
solute carrier family 41 (magnesium transporter), member 1
Gene Aliases
-
Chromosome Location
Chr.13: 48607308 - 48627999 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108855.1 NP_001102325.1 3 - 4 1508 83
XM_008769486.1 XP_008767708.1 2 - 3 631 83

View Full Product Details