Assay Design



Gene Transcripts

Gene Symbol
Spc25
Entrez Gene ID
295661
Gene Name
SPC25, NDC80 kinetochore complex component
Gene Aliases
RGD1307811, Spbc25
Chromosome Location
Chr.3: 55438534 - 55451934 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001009654.1 NP_001009654.1 5 - 6 519 100
XM_006234288.2 XP_006234350.1 5 - 6 525 100
GenBank mRNA BC088213.1 - 5 - 6 519 100

View Full Product Details