Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031793.1 | NP_113981.1 | 12 - 13 | 1263 | 125 |
XM_006234324.2 | XP_006234386.1 | 12 - 13 | 1639 | 125 | |
XM_006234325.2 | XP_006234387.1 | 12 - 13 | 1460 | 125 | |
XM_006234326.2 | XP_006234388.1 | 12 - 13 | 1583 | 125 | |
XM_006234327.2 | XP_006234389.1 | 11 - 12 | 1535 | 125 | |
XM_006234328.2 | XP_006234390.1 | 12 - 13 | 1530 | 125 | |
XM_006234330.2 | XP_006234392.1 | 11 - 12 | 1356 | 125 | |
XM_006234331.2 | XP_006234393.1 | 11 - 12 | 1482 | 125 | |
XM_006234333.2 | XP_006234395.1 | 11 - 12 | 1496 | 125 | |
XM_006234334.2 | XP_006234396.1 | 11 - 12 | 1317 | 125 | |
XM_006234335.2 | XP_006234397.1 | 11 - 12 | 1443 | 125 | |
XM_008761930.1 | XP_008760152.1 | 11 - 12 | 1559 | 125 | |
XM_008761931.1 | XP_008760153.1 | 11 - 12 | 1520 | 125 | |
GenBank mRNA | AF043642.1 | - | 12 - 13 | 1263 | 125 |
FQ230786.1 | - | 7 - 8 | 778 | 125 | |
FQ232477.1 | - | 7 - 8 | 779 | 125 |