Assay Design



Gene Transcripts

Gene Symbol
Nup35
Entrez Gene ID
295692
Gene Name
nucleoporin 35
Gene Aliases
-
Chromosome Location
Chr.3: 67866830 - 67896828 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004229.1 NP_001004229.1 4 - 5 426 81
XM_006234430.2 XP_006234492.1 4 - 5 433 81
GenBank mRNA BC078914.1 - 4 - 5 426 81

View Full Product Details