Assay Design
Gene Transcripts
- Gene Symbol
- Oip5
- Entrez Gene ID
-
499873
- Gene Name
- Opa interacting protein 5
- Gene Aliases
-
RGD1564263
- Chromosome Location
-
Chr.3: 111413662 - 111422319 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC167069.1 |
- |
2 - 3 |
375 |
69 |
View Full Product Details