Assay Design



Gene Transcripts

Gene Symbol
Fbxo3
Entrez Gene ID
690634
Gene Name
F-box protein 3
Gene Aliases
-
Chromosome Location
Chr.3: 93968830 - 94000863 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109606.1 NP_001103076.1 5 - 6 700 82
XM_006234656.2 XP_006234718.1 5 - 6 725 82
XM_006234657.2 XP_006234719.1 5 - 6 725 82

View Full Product Details