Assay Design



Gene Transcripts

Gene Symbol
Scn3a
Entrez Gene ID
497770
Gene Name
sodium channel, voltage-gated, type III, alpha subunit
Gene Aliases
Nav1.3, SCIII, Scn2a
Chromosome Location
Chr.3: 51530897 - 51643140 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013119.1 NP_037251.1 12 - 13 2291 99
XM_008761919.1 XP_008760141.1 11 - 12 2094 99
XM_008761920.1 XP_008760142.1 11 - 12 2094 99
GenBank mRNA Y00766.1 - 12 - 13 2291 99

View Full Product Details