Assay Design



Gene Transcripts

Gene Symbol
Serinc4
Entrez Gene ID
311358
Gene Name
serine incorporator 4
Gene Aliases
RGD1566077
Chromosome Location
Chr.3: 113417429 - 113423555 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001110811.1 NP_001104281.1 2 - 3 591 84
XM_006234853.2 XP_006234915.1 2 - 3 673 84

View Full Product Details