Assay Design



Gene Transcripts

Gene Symbol
Ciao1
Entrez Gene ID
29231
Gene Name
cytosolic iron-sulfur assembly component 1
Gene Aliases
Wdr39
Chromosome Location
Chr.3: 119671778 - 119677324 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008766.1 NP_001008766.1 5 - 6 770 87
GenBank mRNA BC088474.1 - 5 - 6 770 87
FQ230934.1 - 5 - 6 784 87
FQ231672.1 - 5 - 6 763 88

View Full Product Details