Assay Design



Gene Transcripts

Gene Symbol
Nutm1
Entrez Gene ID
366153
Gene Name
NUT midline carcinoma, family member 1
Gene Aliases
Nut, RGD1564529
Chromosome Location
Chr.3: 103733935 - 103745236 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108957.1 NP_001102427.1 2 - 3 833 81
XM_006234708.2 XP_006234770.1 2 - 3 829 81

View Full Product Details