Assay Design



Gene Transcripts

Gene Symbol
Cry2
Entrez Gene ID
170917
Gene Name
cryptochrome circadian clock 2
Gene Aliases
-
Chromosome Location
Chr.3: 81314151 - 81344109 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133405.2 NP_596896.2 11 - 12 1795 65
GenBank mRNA BC166416.1 - 11 - 12 1795 65
FQ211347.1 - 11 - 12 1805 65

View Full Product Details