Assay Design



Gene Transcripts

Gene Symbol
Dnajc24
Entrez Gene ID
362184
Gene Name
DnaJ (Hsp40) homolog, subfamily C, member 24
Gene Aliases
Dph4, RGD1564710, Zcsl3
Chromosome Location
Chr.3: 96025388 - 96065744 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191853.1 NP_001178782.1 1 - 2 43 77
XM_006234654.2 XP_006234716.1 1 - 2 76 77

View Full Product Details