Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001037358.2 | NP_001032435.2 | 3 - 4 | 661 | 78 |
NM_001244779.1 | NP_001231708.1 | 3 - 4 | 551 | 78 | |
NM_001244780.1 | NP_001231709.1 | 3 - 4 | 526 | 78 | |
NM_001244781.1 | NP_001231710.1 | 3 - 4 | 556 | 78 | |
GenBank mRNA | BC105772.1 | - | 3 - 4 | 661 | 78 |
CK476274.1 | - | 3 - 4 | 512 | 78 | |
CO559731.1 | - | - | 526 | 78 | |
DY319476.1 | - | 3 - 4 | 552 | 78 |