Assay Design



Gene Transcripts

Gene Symbol
Mmp12
Entrez Gene ID
117033
Gene Name
matrix metallopeptidase 12
Gene Aliases
Mme
Chromosome Location
Chr.8: 5606592 - 5616494 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053963.2 NP_446415.2 3 - 4 532 111
GenBank mRNA BC088120.1 - 3 - 4 532 111
X98517.1 - 3 - 4 532 111

View Full Product Details