Assay Design



Gene Transcripts

Gene Symbol
Rp9
Entrez Gene ID
363032
Gene Name
retinitis pigmentosa 9 (human)
Gene Aliases
RGD1559759, Rp9h
Chromosome Location
Chr.8: 23450111 - 23485759 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008765960.1 XP_008764182.1 4 - 5 596 145

View Full Product Details