Assay Design



Gene Transcripts

Gene Symbol
Ldlr
Entrez Gene ID
300438
Gene Name
low density lipoprotein receptor
Gene Aliases
LDLRA
Chromosome Location
Chr.8: 22750425 - 22773305 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_175762.2 NP_786938.1 6 - 7 1092 71
GenBank mRNA X13722.1 - 6 - 7 1092 71

View Full Product Details