Assay Design



Gene Transcripts

Gene Symbol
Slc36a4
Entrez Gene ID
315439
Gene Name
solute carrier family 36 (proton/amino acid symporter), member 4
Gene Aliases
-
Chromosome Location
Chr.8: 14238971 - 14271931 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108127.1 NP_001101597.1 7 - 8 786 74
XM_006242558.2 XP_006242620.1 7 - 8 1409 74
XM_008765939.1 XP_008764161.1 4 - 5 471 74

View Full Product Details