Assay Design



Gene Transcripts

Gene Symbol
Ap1m2
Entrez Gene ID
367038
Gene Name
adaptor-related protein complex 1, mu 2 subunit
Gene Aliases
RGD1561490
Chromosome Location
Chr.8: 22318941 - 22336827 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108996.2 NP_001102466.1 6 - 7 705 65
GenBank mRNA BC167082.1 - 6 - 7 705 65

View Full Product Details