Assay Design



Gene Transcripts

Gene Symbol
Tmed1
Entrez Gene ID
315461
Gene Name
transmembrane emp24 protein transport domain containing 1
Gene Aliases
Il1rl1l
Chromosome Location
Chr.8: 22540422 - 22544371 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013432.1 NP_001013450.1 2 - 3 288 61
XM_006242644.1 XP_006242706.1 3 - 4 1767 61
XM_008765940.1 XP_008764162.1 3 - 4 1702 61
GenBank mRNA BC091167.1 - 2 - 3 288 61

View Full Product Details