Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053736.2 | NP_446188.2 | 7 - 8 | 1138 | 158 |
GenBank mRNA | BC104720.1 | - | 7 - 8 | 1138 | 158 |
FQ227402.1 | - | 7 - 8 | 1175 | 158 | |
FQ232279.1 | - | 7 - 8 | 1175 | 158 |