Assay Design



Gene Transcripts

Gene Symbol
Rsu1
Entrez Gene ID
680419
Gene Name
Ras suppressor protein 1
Gene Aliases
-
Chromosome Location
Chr.17: 80388477 - 80577338 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109404.1 NP_001102874.1 - 248 84

View Full Product Details