Assay Design



Gene Transcripts

Gene Symbol
Ucma
Entrez Gene ID
291312
Gene Name
upper zone of growth plate and cartilage matrix associated
Gene Aliases
GRP, RGD1308977
Chromosome Location
Chr.17: 77252099 - 77262296 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106121.1 NP_001099591.1 2 - 3 280 55
XM_006254265.1 XP_006254327.1 4 - 5 920 55
GenBank mRNA EU022754.1 - 4 - 5 355 55

View Full Product Details