Assay Design



Gene Transcripts

Gene Symbol
Phyh
Entrez Gene ID
114209
Gene Name
phytanoyl-CoA 2-hydroxylase
Gene Aliases
-
Chromosome Location
Chr.17: 77287580 - 77304482 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053674.1 NP_446126.1 3 - 4 255 121
GenBank mRNA AF121345.1 - 3 - 4 255 121
BC086573.1 - 3 - 4 250 121

View Full Product Details