Assay Design



Gene Transcripts

Gene Symbol
Spag6l
Entrez Gene ID
291340
Gene Name
sperm associated antigen 6-like
Gene Aliases
Spag6
Chromosome Location
Chr.17: 85377769 - 85437995 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106125.1 NP_001099595.1 6 - 7 855 78
XM_006254307.2 XP_006254369.1 7 - 8 4769 78

View Full Product Details