Assay Design
Gene Transcripts
- Gene Symbol
- Prpf18
- Entrez Gene ID
-
171552
- Gene Name
- pre-mRNA processing factor 18
- Gene Aliases
-
Pprf18
- Chromosome Location
-
Chr.17: 77601914 - 77632507 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_138523.2 |
NP_612532.1 |
7 - 8 |
821 |
106 |
GenBank mRNA |
AF244920.1 |
- |
7 - 8 |
816 |
106 |
View Full Product Details