Assay Design



Gene Transcripts

Gene Symbol
Aga
Entrez Gene ID
290923
Gene Name
aspartylglucosaminidase
Gene Aliases
-
Chromosome Location
Chr.16: 41222225 - 41234163 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031641.1 NP_001026811.1 6 - 7 698 72
XM_006253111.2 XP_006253173.1 7 - 8 823 72
GenBank mRNA BC098718.1 - 6 - 7 698 72

View Full Product Details