Assay Design



Gene Transcripts

Gene Symbol
Asb5
Entrez Gene ID
361187
Gene Name
ankyrin repeat and SOCS box-containing 5
Gene Aliases
-
Chromosome Location
Chr.16: 39985233 - 40036284 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001044247.1 NP_001037712.1 4 - 5 598 89
XM_006253100.2 XP_006253162.1 4 - 5 2028 89
XM_008771246.1 XP_008769468.1 4 - 5 1031 89
XM_008771247.1 XP_008769469.1 4 - 5 626 89
XM_008771248.1 XP_008769470.1 4 - 5 1871 89
GenBank mRNA BC098935.1 - 4 - 5 598 89
FQ215658.1 - 4 - 5 470 89

View Full Product Details