Assay Design



Gene Transcripts

Gene Symbol
Irf2
Entrez Gene ID
290749
Gene Name
interferon regulatory factor 2
Gene Aliases
-
Chromosome Location
Chr.16: 48582354 - 48732129 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001047086.1 NP_001040551.1 2 - 3 205 100
XM_006253139.2 XP_006253201.1 2 - 3 466 100
XM_008771256.1 XP_008769478.1 2 - 3 301 100
GenBank mRNA BC166593.1 - 2 - 3 246 100
DQ674266.1 - 2 - 3 205 100

View Full Product Details