Assay Design



Gene Transcripts

Gene Symbol
Rwdd4
Entrez Gene ID
502084
Gene Name
RWD domain containing 4A
Gene Aliases
Rwdd4a
Chromosome Location
Chr.16: 47856522 - 47874856 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034994.1 NP_001030166.1 5 - 6 693 109
GenBank mRNA BC092581.1 - 5 - 6 693 109
FQ229862.1 - 5 - 6 734 109

View Full Product Details