Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001013882.2 | NP_001013904.2 | 3 - 4 | 458 | 87 |
GenBank mRNA | BC087138.1 | - | 3 - 4 | 413 | 87 |
DY317307.1 | - | 3 - 4 | 426 | 87 | |
EV776012.1 | - | 3 - 4 | 483 | 87 |