Assay Design



Gene Transcripts

Gene Symbol
Zdhhc20
Entrez Gene ID
305923
Gene Name
zinc finger, DHHC-type containing 20
Gene Aliases
RGD1305755
Chromosome Location
Chr.15: 38108886 - 38165987 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039336.1 NP_001034425.1 9 - 10 1039 92
XM_006252072.2 XP_006252134.1 10 - 11 1137 92
XM_006252073.2 XP_006252135.1 10 - 11 1337 92
XM_008770749.1 XP_008768971.1 10 - 11 1152 92
XM_008770750.1 XP_008768972.1 10 - 11 1338 92
GenBank mRNA AY886539.1 - 9 - 10 1039 92
BC098018.1 - 9 - 10 1015 92
BC127440.1 - 9 - 10 1061 92

View Full Product Details