Assay Design



Gene Transcripts

Gene Symbol
Khnyn
Entrez Gene ID
498517
Gene Name
KH and NYN domain containing
Gene Aliases
Khnyn-ps1, RGD1565688
Chromosome Location
Chr.15: 34551573 - 34565410 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003751468.3 XP_003751516.1 6 - 7 1921 72
XM_006252032.2 XP_006252094.1 6 - 7 2116 72

View Full Product Details